Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETHE1 Mouse anti-Human, Mouse, Rat, FITC, Clone: 1621, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243632F

Catalog No. NBP243632F


Add to cart

Description

Description

ETHE1 Monoclonal antibody specifically detects ETHE1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ETHE1
1621
Immunocytochemistry/Immunofluorescence
Purified
ETHE1
Full length human ETHE1 Recombinant protein.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12
Mouse
IgG2b
0.1 ml
Primary
23474
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit