Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Anhydrous, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry, bulk pharmaceutical chemical
Flash Point −45.00°C (113°F)
Specific Gravity 1L = 0.71kg
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Quantity 4L
Formula Weight 74.12
Grade Meets USP requirements
Chemical Name or Material Ether, Anhydrous