Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ether, Anhydrous, HPLC Grade, J.T. Baker™
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
Description
- For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry, bulk pharmaceutical chemical
Specifications
Flash Point | −45.00°C (113°F) |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 4 L |
Specific Gravity | 1L = 0.71kg |
Formula Weight | 74.12 |
Grade | Meets USP requirements |
Chemical Name or Material | Ether, Anhydrous |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction