Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Anhydrous, min. 99.0%, BAKER ANALYZED™ ACS Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

High quality chemicals for laboratory and specialized industrial use

    Bulk Pharmaceutical Chemical
Density 0.7079g/mL
Packaging Aluminum Bottle
Flash Point −45°C (− 113°C)
Specific Gravity 1L = 0.71kg
Quantity 1L
Residue after Evaporation max. 0.001%
Physical Form Liquid
Percent Purity ≥99.0%
Grade Meets USP requirements
Chemical Name or Material Anhydrous, Contains BHT as a Preservative

Recommended Storage : Keep in partly filled, tightly closed, light resistant containers, at a temperature not exceeding 30°C (86°F), remote from fire