Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ether, Anhydrous, min. 99.0%, BAKER ANALYZED™ ACS Reagent, J.T. Baker™
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Description
- Bulk Pharmaceutical Chemical
Specifications
Density | 0.7079g/mL |
Flash Point | −45°C (− 113°C) |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 4 L |
Residue after Evaporation | max. 0.001% |
Specific Gravity | 1L = 0.71kg |
Percent Purity | ≥99.0% |
Grade | Meets USP requirements |
Physical Form | Liquid |
Chemical Name or Material | Anhydrous, Contains BHT as a Preservative |
Safety and Handling
Recommended Storage | Keep in partly filled, tightly closed, light resistant containers, at a temperature not exceeding 30°C (86°F), remote from fire |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction