Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether ChromAR, Macron Fine Chemicals™

Click to view available options
Quantity:
1L
3.75L
Packaging:
Al Bottle
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
Formula Weight 74.12
Packaging Al Bottle
Quantity 3.75L
Chemical Name or Material Ether