Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Contains Alcohol, Water, and BHT as Preservatives, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
CHEBI:35702

High quality chemicals for laboratory and specialized industrial use

SMILES CCOCC
IUPAC Name ethoxyethane
Formula Weight 74.12
Packaging Aluminum Bottle
Specific Gravity 1L = 0.71kg
Quantity 4L
Flash Point −45.00°C
Chemical Name or Material Ether, Contains Alcohol, Water, and BHT as Preservatives
Grade Baker Analyzed™ ACS Reagent