Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Contains Alcohol, Water, and BHT as Preservatives, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Flash Point −45.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Ether, Contains Alcohol, Water, and BHT as Preservatives