Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ether, Contains Alcohol, Water, and BHT as Preservatives, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Specifications
Specifications
Flash Point | −45.00°C |
Packaging | Aluminum Bottle |
Quantity | 4 L |
Specific Gravity | 1L = 0.71kg |
Formula Weight | 74.12 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Ether, Contains Alcohol, Water, and BHT as Preservatives |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction