Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Preserved with 2% Alcohol, 0.5% max Water and 7ppm min BHT, U.S.P., J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Aluminum Bottle

Chemical Identifiers

60-29-7
C4H10O
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
CHEBI:35702
ethoxyethane
CCOCC
Formula Weight 74.12
Packaging Aluminum Bottle
Specific Gravity 1L = 0.71kg
Quantity 1L
Flash Point −45.00°C
Chemical Name or Material Ether, Preserved with 2% Alcohol, 0.5% max Water and 7ppm min BHT
Grade USP