Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ether, Stabilized with Ethanol, SpectroSolv (TM), 99%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
60-29-7
60-29-7
CAS Max % 1
Percent Purity 0.99
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Stabilizer 1.5 - 2.5%
Titratable Acid 0.2 μeq/g
Water 0.0003
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.