Available on GSA/VA Contract for Federal Government customers only. Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents

$71.92

Chemical Identifiers

Chemical Name or Material Ethidium Bromide
Name Note 1% Solution
CAS [1239-45-8], [7732-18-5]
Purity Grade Molecular Biology-Grade
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Reddish-Brown Liquid
Molecular Formula C21H20N3Br
Formula Weight 394.33
UN Number 2810
Merck Index 15, 4769
View More Specs

  • Products 1
  • Description & Specifications
  • Safety and Handling
Products
Catalog Number Description Price Quantity Availability  

BP1302-10

Available on GSA/VA Contract for Federal Government customers only.  SDS
Amber Poly Bottle; 10mL
Each for $71.92
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products