Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethidium Bromide, BiotechGrade, 98%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
1239-45-8
C21H20BrN3
394.32
ZMMJGEGLRURXTF-UHFFFAOYSA-N
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
1239-45-8
C21H20BrN3
394.32
ZMMJGEGLRURXTF-UHFFFAOYSA-N
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

E1018, 1239-45-8, C21H20BrN3

Spectrum™ Ethidium Bromide, BiotechGrade , also known as 2,7-Diamino-10-ethyl-6-phenylphenanthridinium bromide, is an intercalating agent often used in molecular biology laboratories as a nucleic acid stain for techniques like as agarose gel electrophoresis. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Packaging Amber Glass Bottle
Quantity 1 g
CAS Max % 1
UN Number UN2811
Percent Purity 98%