Learn More
Ethilon Nylon Precision Point-Reverse Cutting Sutures
A nonabsorbable, sterile, surgical, monofilament suture composed of the long-chain, aliphatic polymers Nylon 6 and Nylon 6.6
Manufacturer:  Moore Medical Corp 47971
 View more versions of this product
Catalog No. 19-898-172
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
18 in.
Aliphatic Polymers Nylon 6 and Nylon 6.6
12 Box
Black
1.3cm
3/8 Circle
45cm
Suture
Yes
0.511 in.
PS-3
6 to 0