Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethionamide, 98%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
536-33-4
C8H10N2S
166.242
MFCD00057361
AEOCXXJPGCBFJA-UHFFFAOYSA-N
2-Ethyl-4-pyridinecarbothioamide
2761171
CHEBI:4885
2-ethylpyridine-4-carbothioamide
CCC1=NC=CC(=C1)C(=S)N
536-33-4
C8H10N2S
166.242
MFCD00057361
AEOCXXJPGCBFJA-UHFFFAOYSA-N
2-Ethyl-4-pyridinecarbothioamide
2761171
CHEBI:4885
2-ethylpyridine-4-carbothioamide
CCC1=NC=CC(=C1)C(=S)N
Packaging Plastic bottle
Formula Weight 166.24
Physical Form Powder
Percent Purity 98%
Boiling Point 297°C
Melting Point 160°C to 162°C (decomposition)
Quantity 100g
Flash Point >110°C
Chemical Name or Material Ethionamide