Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethosuximide, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
77-67-8
C7H11NO2
141.17
HAPOVYFOVVWLRS-UHFFFAOYNA-N
3-ethyl-3-methylpyrrolidine-2,5-dione
CCC1(C)CC(=O)NC1=O
77-67-8
C7H11NO2
141.17
HAPOVYFOVVWLRS-UHFFFAOYNA-N
3-ethyl-3-methylpyrrolidine-2,5-dione
CCC1(C)CC(=O)NC1=O

E2017, 77-67-8, C7H11NO2

Spectrum™ Ethosuximide, USP , a succinimide anticonvulsant, is used in absence seizures as well as for intermittent explosive disorder. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Water 0.005
Ignition Residue 0.005
Melting Range 47-52° C
Quantity 5 kg
CAS Max % 1
Percent Purity 98 to 101%