Alfa Aesar™ Ethoxyacetic acid, 98%

Manufacturer: Alfa Aesar™ B2419414

Quantity 25g

 View more versions of this product

Catalog No. AAB2419414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 627-03-2
    Molecular Formula CH3CH2OCH2CO2H
    Formula Weight 104.11
    MDL Number MFCD00004310
    UN Number UN3265
    Beilstein 1743037
    Chemical Name or Material Ethoxyacetic acid

    Specifications

    Formula Weight 104.11
    TSCA TSCA
    Molecular Formula CH3CH2OCH2CO2H
    UN Number UN3265
    Boiling Point 207°C
    Odor Pungent
    RTECS Number AH3600000
    MDL Number MFCD00004310
    CAS 627-03-2
    Beilstein 1743037
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    Flash Point 97°C (206°F)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Ethoxyacetic acid
    Assay 98%

    Description

    Applications
    Ethoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.

    Solubility
    Miscible with water.

    Notes
    Incompatible with bases, oxidizing agents and reducing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit