Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethoxycarbonyl isothiocyanate, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
16182-04-0
C4H5NO2S
131.15
MFCD00004814
BDTDECDAHYOJRO-UHFFFAOYSA-N
ethoxycarbonyl isothiocyanate,ethyl isothiocyanatoformate,carbethoxy isothiocyanate,ethoxycarbonylisothiocyanate,ethyl n-carbothioylcarbamate,ethyl n-sulfanylidenemethylidene carbamate,ethyl isothiocyanatidocarbonate,carbon isothiocyanatidic acid, ethyl ester,o-ethyl carbonisothiocyanatidate
85320
ethyl N-(sulfanylidenemethylidene)carbamate
CCOC(=O)N=C=S
16182-04-0
C4H5NO2S
131.15
MFCD00004814
BDTDECDAHYOJRO-UHFFFAOYSA-N
ethoxycarbonyl isothiocyanate,ethyl isothiocyanatoformate,carbethoxy isothiocyanate,ethoxycarbonylisothiocyanate,ethyl n-carbothioylcarbamate,ethyl n-sulfanylidenemethylidene carbamate,ethyl isothiocyanatidocarbonate,carbon isothiocyanatidic acid, ethyl ester,o-ethyl carbonisothiocyanatidate
85320
ethyl N-(sulfanylidenemethylidene)carbamate
CCOC(=O)N=C=S
Flash Point 50°C
Boiling Point 56.0°C (18.0 mmHg)
Density 1.1600g/mL
Specific Gravity 1.16
Quantity 25g
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Linear Formula SCNCOOCH2CH3
Beilstein 03, III, 279
Fieser 04,223; 06,250; 07,146
Formula Weight 131.15
Percent Purity 95%
Chemical Name or Material Ethoxycarbonyl isothiocyanate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Danger

EINECSNumber : 240-318-9