Alfa Aesar™ Ethyl 1,1,2,2-tetrafluoroethyl ether, 97%

Manufacturer: Alfa Aesar™ B2270306

Quantity 5g

 View more versions of this product

Catalog No. AAB2270306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 512-51-6
    Molecular Formula C4H6F4O
    Linear Formula HCF2CF2OCH2CH3
    Formula Weight 146.08
    MDL Number MFCD00069162
    UN Number UN3271
    Chemical Name or Material Ethyl 1,1,2,2-tetrafluoroethyl ether

    Specifications

    Formula Weight 146.08
    Molecular Formula C4H6F4O
    Boiling Point 56° to 58°C
    Quantity 5g
    Flash Point −15°C (−5°F)
    Linear Formula HCF2CF2OCH2CH3
    MDL Number MFCD00069162
    CAS 512-51-6
    UN Number UN3271
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    Chemical Name or Material Ethyl 1,1,2,2-tetrafluoroethyl ether
    Assay 97%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit