Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, 95%

Click to view available options
Quantity:
250mg
1g
5g
41234-43-9
C12H15NO2
205.257
MFCD04035517
RGHMPTHWVVRXHW-UHFFFAOYSA-N
1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid ethyl ester,1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid ethyl ester,ethyl 1,2,3,4-tetrahydro-3-isoquinolinecarboxylate,3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, ethyl ester,1,2,3,4-tetrahydroisoquinoline-3-carboxylic ethyl ester,1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester,+/--1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester,1,2,3,4-tetrahydro-3-isoquinoline carboxylic acid, ethyl ester,3-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-, ethyl ester
652898
ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
CCOC(=O)C1CC2=CC=CC=C2CN1
41234-43-9
C12H15NO2
205.257
MFCD04035517
RGHMPTHWVVRXHW-UHFFFAOYSA-N
1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid ethyl ester,1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid ethyl ester,ethyl 1,2,3,4-tetrahydro-3-isoquinolinecarboxylate,3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, ethyl ester,1,2,3,4-tetrahydroisoquinoline-3-carboxylic ethyl ester,1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester,+/--1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ethyl ester,1,2,3,4-tetrahydro-3-isoquinoline carboxylic acid, ethyl ester,3-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-, ethyl ester
652898
ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
CCOC(=O)C1CC2=CC=CC=C2CN1
Quantity 250mg
Formula Weight 205.26
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate