Alfa Aesar™ Ethyl 1-benzylpiperidine-4-carboxylate, 97+%

Manufacturer: Alfa Aesar™ H52374MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52374MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24228-40-8
    Molecular Formula C15H21NO2
    Formula Weight 247.33
    MDL Number MFCD00040748
    Synonym 1-Benzylpiperidine-4-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 1-benzylpiperidine-4-carboxylate

    Specifications

    Formula Weight 247.33
    Molecular Formula C15H21NO2
    MDL Number MFCD00040748
    CAS 24228-40-8
    Synonym 1-Benzylpiperidine-4-carboxylic acid ethyl ester
    Quantity 250mg
    Chemical Name or Material Ethyl 1-benzylpiperidine-4-carboxylate
    Assay ≥97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit