Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 1-benzylpyrrolidine-3-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
25g
5747-92-2
C14H19NO2
233.311
MFCD04039852
CYPXEPWPTXKUPL-UHFFFAOYSA-N
1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester,ethyl 1-benzyl-pyrrolidine-3-carboxylate,3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester,ethyl1-benzylpyrrolidine-3-carboxylate,1-benzylpyrrolidine-3-carboxylic acid ethyl ester,acmc-1asr8,1-benzyl-pyrrolidine-3-carboxylicacidethylester,ksc495s0t,1-benzyl-3-carboethoxypyrrolidine,ethyl 1-benzyl pyrrolidine-3-carboxylate
11622765
ethyl 1-benzylpyrrolidine-3-carboxylate
CCOC(=O)C1CCN(C1)CC2=CC=CC=C2
5747-92-2
C14H19NO2
233.311
MFCD04039852
CYPXEPWPTXKUPL-UHFFFAOYSA-N
1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester,ethyl 1-benzyl-pyrrolidine-3-carboxylate,3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester,ethyl1-benzylpyrrolidine-3-carboxylate,1-benzylpyrrolidine-3-carboxylic acid ethyl ester,acmc-1asr8,1-benzyl-pyrrolidine-3-carboxylicacidethylester,ksc495s0t,1-benzyl-3-carboethoxypyrrolidine,ethyl 1-benzyl pyrrolidine-3-carboxylate
11622765
ethyl 1-benzylpyrrolidine-3-carboxylate
CCOC(=O)C1CCN(C1)CC2=CC=CC=C2
Quantity 5g
Color Yellow
Beilstein 398388
Formula Weight 233.31
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 1-benzylpyrrolidine-3-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning