Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-benzylpyrrolidine-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number AAH6496203
Click to view available options
Quantity:
1 g
5 g
25 g
5747-92-2
C14H19NO2
233.311
MFCD04039852
CYPXEPWPTXKUPL-UHFFFAOYSA-N
1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 1-benzyl-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, ethyl1-benzylpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid ethyl ester, acmc-1asr8, 1-benzyl-pyrrolidine-3-carboxylicacidethylester, ksc495s0t, 1-benzyl-3-carboethoxypyrrolidine, ethyl 1-benzyl pyrrolidine-3-carboxylate
11622765
ethyl 1-benzylpyrrolidine-3-carboxylate
CCOC(=O)C1CCN(C1)CC2=CC=CC=C2
5747-92-2
C14H19NO2
233.311
MFCD04039852
CYPXEPWPTXKUPL-UHFFFAOYSA-N
1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 1-benzyl-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, ethyl1-benzylpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid ethyl ester, acmc-1asr8, 1-benzyl-pyrrolidine-3-carboxylicacidethylester, ksc495s0t, 1-benzyl-3-carboethoxypyrrolidine, ethyl 1-benzyl pyrrolidine-3-carboxylate
11622765
ethyl 1-benzylpyrrolidine-3-carboxylate
CCOC(=O)C1CCN(C1)CC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 g
Beilstein 398388
Formula Weight 233.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 1-benzylpyrrolidine-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit