Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%

Click to view available options
Quantity:
250mg
1g
5g
71233-25-5
C13H21NO5
271.313
MFCD09878815
WCTXJAXKORIYNA-UHFFFAOYSA-N
1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate,ethyl 1-n-boc-3-oxopiperidine-4-carboxylate,ethyl n-boc-3-oxopiperidine-4-carboxylate,3-oxo-piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester,ethyl 1-n-boc-3-oxo-piperidine-4-carboxylate,ethyl 1-boc-3-oxopiperidine-4-carboxylate,ethyl n-tert-butoxycarbonyl-3-oxo-piperidine-4-carboxylate,1-tert-butyl 4-ethyl 3-oxo-1,4-piperidinedicarboxylate,1,4-piperidinedicarboxylic acid, 3-oxo-, 1-1,1-dimethylethyl 4-ethyl ester,ksc915i7p
15852989
1-O-tert-butyl 4-O-ethyl 3-oxopiperidine-1,4-dicarboxylate
CCOC(=O)C1CCN(CC1=O)C(=O)OC(C)(C)C
71233-25-5
C13H21NO5
271.313
MFCD09878815
WCTXJAXKORIYNA-UHFFFAOYSA-N
1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate,ethyl 1-n-boc-3-oxopiperidine-4-carboxylate,ethyl n-boc-3-oxopiperidine-4-carboxylate,3-oxo-piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester,ethyl 1-n-boc-3-oxo-piperidine-4-carboxylate,ethyl 1-boc-3-oxopiperidine-4-carboxylate,ethyl n-tert-butoxycarbonyl-3-oxo-piperidine-4-carboxylate,1-tert-butyl 4-ethyl 3-oxo-1,4-piperidinedicarboxylate,1,4-piperidinedicarboxylic acid, 3-oxo-, 1-1,1-dimethylethyl 4-ethyl ester,ksc915i7p
15852989
1-O-tert-butyl 4-O-ethyl 3-oxopiperidine-1,4-dicarboxylate
CCOC(=O)C1CCN(CC1=O)C(=O)OC(C)(C)C
Quantity 250mg
Beilstein 421506
Formula Weight 271.31
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 1-Boc-3-oxopiperidine-4-carboxylate

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures