Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-bromocyclobutanecarboxylate, 96%, Thermo Scientific Chemicals

Catalog Number AC188710050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
35120-18-4
C7H11BrO2
207.06
MFCD00001322
UTVNSHXHFRIXMM-UHFFFAOYSA-N
ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane
97595
ethyl 1-bromocyclobutane-1-carboxylate
CCOC(=O)C1(CCC1)Br
35120-18-4
C7H11BrO2
207.06
MFCD00001322
UTVNSHXHFRIXMM-UHFFFAOYSA-N
ethyl 1-bromocyclobutanecarboxylate, ethyl alpha-bromocyclobutanecarboxylate, 1-bromo-cyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 1-bromo-, ethyl ester, 1-bromocyclobutanecarboxylic acid ethyl ester, ethyl .alpha.-bromocyclobutanecarboxylate, acmc-1cswe, ethyl abromocyclobutanecarboxylate, ethyl-1-bromocyclobutanecarboxylate, 1-ethoxycarbonyl-1-bromocyclobutane
97595
ethyl 1-bromocyclobutane-1-carboxylate
CCOC(=O)C1(CCC1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.2790g/mL
Boiling Point 85°C to 88°C (12.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula BrC4H6CO2C2H5
Quantity 5 g
Specific Gravity 1.279
Formula Weight 207.06
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl 1-bromocyclobutanecarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 252-384-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit