Alfa Aesar™ Ethyl(1-hexadecyl)dimethylammonium bromide, 99%

Manufacturer: Alfa Aesar™ B2399122

Quantity 100g

 View more versions of this product

Catalog No. AAB2399122

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 124-03-8
    Molecular Formula CH3(CH2)15N(CH3)2(CH2CH3)Br
    Formula Weight 378.49
    MDL Number MFCD00011774
    Beilstein 3729134
    Merck Index 14,2029
    Synonym Cetyl ethyl dimethylammonium bromide
    Chemical Name or Material Ethyl(1-hexadecyl)dimethylammonium bromide

    Specifications

    Formula Weight 378.49
    Melting Point ≈192°C (dec.)
    TSCA TSCA
    Molecular Formula CH3(CH2)15N(CH3)2(CH2CH3)Br
    MDL Number MFCD00011774
    Beilstein 3729134
    Merck Index 14,2029
    Synonym Cetyl ethyl dimethylammonium bromide
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 100g
    RTECS Number BQ5425000
    CAS 124-03-8
    Health Hazard 1 Warning
    Chemical Name or Material Ethyl(1-hexadecyl)dimethylammonium bromide
    Assay 99%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit