Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-methylnipecotate, +98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5166-67-6
C9H17NO2
171.24
MFCD00006496
VFJJNMLPRDRTCO-UHFFFAOYSA-N
ethyl 1-methylnipecotate,ethyl n-methylpiperidine-3-carboxylate,ethyl 1-methyl-3-piperidinecarboxylate,1-methyl-piperidine-3-carboxylic acid ethyl ester,1-methylpiperidine-3-carboxylic acid ethyl ester,1-methyl-3-piperidinecarboxylic acid ethyl ester,3-piperidinecarboxylic acid, 1-methyl-, ethyl ester,ethyl-1-methylpiperidin-3-carboxylat,pubchem11774,acmc-209kvb
97981
ethyl 1-methylpiperidine-3-carboxylate
CCOC(=O)C1CCCN(C1)C
5166-67-6
C9H17NO2
171.24
MFCD00006496
VFJJNMLPRDRTCO-UHFFFAOYSA-N
ethyl 1-methylnipecotate,ethyl n-methylpiperidine-3-carboxylate,ethyl 1-methyl-3-piperidinecarboxylate,1-methyl-piperidine-3-carboxylic acid ethyl ester,1-methylpiperidine-3-carboxylic acid ethyl ester,1-methyl-3-piperidinecarboxylic acid ethyl ester,3-piperidinecarboxylic acid, 1-methyl-, ethyl ester,ethyl-1-methylpiperidin-3-carboxylat,pubchem11774,acmc-209kvb
97981
ethyl 1-methylpiperidine-3-carboxylate
CCOC(=O)C1CCCN(C1)C
Quantity 5g
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4490 to 1.4510 (20°C, 589nm)
Density 0.9540g/mL
Assay Percent Range 98+%
Beilstein 22, V,1, 237
Formula Weight 171.24
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material Ethyl 1-methylnipecotate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.