Alfa Aesar™ Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%

Manufacturer: Alfa Aesar™ H32847MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32847MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H11F3O2
    Formula Weight 196.17
    MDL Number MFCD10699116
    Synonym 1-(Trifluoromethyl)cyclobutanecarboxylic acid ethyl ester
    Chemical Name or Material Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate

    Specifications

    Formula Weight 196.17
    Molecular Formula C8H11F3O2
    MDL Number MFCD10699116
    Synonym 1-(Trifluoromethyl)cyclobutanecarboxylic acid ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Boiling Point 61° to 63°C (20mmHg)
    Quantity 250mg
    Chemical Name or Material Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit