Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 1H-indazole-3-carboxylate, 95%

Manufacturer:  Alfa Aesar™ H3209803

Quantity 1g

 View more versions of this product

Catalog No. AAH3209803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

4498-68-4
190.2
1H-Indazole-3-carboxylic acid ethyl ester
C10H10N2O2
MFCD01138134
Ethyl 1H-indazole-3-carboxylate

Specifications

4498-68-4
190.2
1H-Indazole-3-carboxylic acid ethyl ester
1g
95%
C10H10N2O2
MFCD01138134
136° to 138°C
Ethyl 1H-indazole-3-carboxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit