Alfa Aesar™ Ethyl 1H-indazole-5-carboxylate, 95%

Manufacturer: Alfa Aesar™ H32649MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32649MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 192944-51-7
    Molecular Formula C10H10N2O2
    Formula Weight 190.2
    MDL Number MFCD03426220
    Synonym 1H-Indazole-5-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 1H-indazole-5-carboxylate

    Specifications

    Formula Weight 190.2
    Molecular Formula C10H10N2O2
    MDL Number MFCD03426220
    Synonym 1H-Indazole-5-carboxylic acid ethyl ester
    Melting Point 122° to 124°C
    Quantity 250mg
    CAS 192944-51-7
    Chemical Name or Material Ethyl 1H-indazole-5-carboxylate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit