Alfa Aesar™ Ethyl 1H-pyrazole-4-carboxylate, 98%

Manufacturer: Alfa Aesar™ B2535103

Quantity 1g

 View more versions of this product

Catalog No. AAB2535103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37622-90-5
    Molecular Formula C6H8N2O2
    Formula Weight 140.14
    MDL Number MFCD00010844
    Synonym 1H-Pyrazole-4-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 1H-pyrazole-4-carboxylate

    Specifications

    Formula Weight 140.14
    Molecular Formula C6H8N2O2
    MDL Number MFCD00010844
    Synonym 1H-Pyrazole-4-carboxylic acid ethyl ester
    Boiling Point 138° to 140°C (3mmHg)
    Melting Point 75° to 80°C
    CAS 37622-90-5
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Chemical Name or Material Ethyl 1H-pyrazole-4-carboxylate
    Assay 98%

    Description

    Applications
    Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

    Solubility
    Miscible with acetone.

    Notes
    Incompatible with strong oxidizing agents, strong acids and bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit