Alfa Aesar™ Ethyl 2,2-difluoropropionate, 97%

Manufacturer: Alfa Aesar™ H3185203

Quantity 1g

 View more versions of this product

Catalog No. AAH3185203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 28781-85-3
    Molecular Formula C5H8F2O2
    Linear Formula CH3CF2CO2CH2CH3
    Formula Weight 138.11
    MDL Number MFCD06657970
    UN Number UN3272
    Synonym 2,2-Difluoropropionic acid ethyl ester
    Chemical Name or Material Ethyl 2,2-difluoropropionate

    Specifications

    Formula Weight 138.11
    Molecular Formula C5H8F2O2
    Linear Formula CH3CF2CO2CH2CH3
    MDL Number MFCD06657970
    CAS 28781-85-3
    UN Number UN3272
    Synonym 2,2-Difluoropropionic acid ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Boiling Point 24°C (20mmHg)
    Quantity 1g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Ethyl 2,2-difluoropropionate
    Assay 97%

    Description

    Applications
    It is used as a pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in cool dry place. Keep away from oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit