Alfa Aesar™ Ethyl 2,3,4,5-tetrafluorobenzoate, 98%

Manufacturer: Alfa Aesar™ B2192009

Quantity 10g

 View more versions of this product

Catalog No. AAB2192009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122894-73-9
    Molecular Formula C9H6F4O2
    Formula Weight 222.14
    MDL Number MFCD02094158
    Synonym 2,3,4,5-Tetrafluorobenzoic acid ethyl ester
    Chemical Name or Material Ethyl 2,3,4,5-tetrafluorobenzoate

    Specifications

    Formula Weight 222.14
    Molecular Formula C9H6F4O2
    MDL Number MFCD02094158
    CAS 122894-73-9
    Synonym 2,3,4,5-Tetrafluorobenzoic acid ethyl ester
    Quantity 10g
    Chemical Name or Material Ethyl 2,3,4,5-tetrafluorobenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit