Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2,3,4,5-tetrafluorobenzoate, 98%

Click to view available options
Quantity:
2g
10g
122894-73-9
C9H6F4O2
222.139
MFCD02094158
SBHMXBYMQZRRLC-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzoic acid ethyl ester,2,3,4,5-tetrafluoro-benzoic acid ethyl ester,pubchem4072,acmc-1c5fu,ethyl2,3,4,5-tetrafluorobenzoate,2,3,4,5-tetrafluoro-benzoicacidethylester,ethyl 2,3,4,5-tetrakis fluoranyl benzoate
2774395
ethyl 2,3,4,5-tetrafluorobenzoate
CCOC(=O)C1=CC(=C(C(=C1F)F)F)F
122894-73-9
C9H6F4O2
222.139
MFCD02094158
SBHMXBYMQZRRLC-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzoic acid ethyl ester,2,3,4,5-tetrafluoro-benzoic acid ethyl ester,pubchem4072,acmc-1c5fu,ethyl2,3,4,5-tetrafluorobenzoate,2,3,4,5-tetrafluoro-benzoicacidethylester,ethyl 2,3,4,5-tetrakis fluoranyl benzoate
2774395
ethyl 2,3,4,5-tetrafluorobenzoate
CCOC(=O)C1=CC(=C(C(=C1F)F)F)F
Quantity 2g
Formula Weight 222.14
Percent Purity 98%
Chemical Name or Material Ethyl 2,3,4,5-tetrafluorobenzoate