Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,3-dibromopropionate, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
3674-13-3
C5H8Br2O2
259.925
MFCD00000212
OENICUBCLXKLJQ-UHFFFAOYSA-N
ethyl 2,3-dibromopropionate,propanoic acid, 2,3-dibromo-, ethyl ester,ethyl alpha,beta-dibromopropionate,propionic acid, 2,3-dibromo-, ethyl ester,ethyl-2,3-dibromopropionate,2,3-dibromopropionic acid ethyl ester,ethyl .alpha.,.beta.-dibromopropionate,propionic acid, 2,3-dibromo-, ethyl ester 6ci,7ci,8ci,ethyl2,3-dibromopropionate,acmc-1ahn6
97945
ethyl 2,3-dibromopropanoate
CCOC(=O)C(CBr)Br
3674-13-3
C5H8Br2O2
259.925
MFCD00000212
OENICUBCLXKLJQ-UHFFFAOYSA-N
ethyl 2,3-dibromopropionate,propanoic acid, 2,3-dibromo-, ethyl ester,ethyl alpha,beta-dibromopropionate,propionic acid, 2,3-dibromo-, ethyl ester,ethyl-2,3-dibromopropionate,2,3-dibromopropionic acid ethyl ester,ethyl .alpha.,.beta.-dibromopropionate,propionic acid, 2,3-dibromo-, ethyl ester 6ci,7ci,8ci,ethyl2,3-dibromopropionate,acmc-1ahn6
97945
ethyl 2,3-dibromopropanoate
CCOC(=O)C(CBr)Br
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 25mL
Density 1.7880g/mL
Refractive Index 1.4976 to 1.5016 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula BrCH2CH(Br)CO2C2H5
Beilstein 02, 259
Formula Weight 259.91
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 2, 3-dibromopropionate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-941-8

RTECSNumber : UA2458382

TSCA : TSCA