Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals

Catalog Number AAH6478806
Click to view available options
Quantity:
250 mg
1 g
5 g
51940-64-8
C7H6Cl2N2O2
221.037
MFCD09910281
SRJBDGLSCPDXBL-UHFFFAOYSA-N
104020
ethyl 2,4-dichloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)Cl
51940-64-8
C7H6Cl2N2O2
221.037
MFCD09910281
SRJBDGLSCPDXBL-UHFFFAOYSA-N
104020
ethyl 2,4-dichloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)Cl

It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 36°C to 37°C
Color White
Quantity 5 g
Beilstein 152173
Synonym ethyl 2,4-dichloro-5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester, ethyl 2,4-dichloro-pyrimidine-5-carboxylate, 2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonylpyrimidine, 2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloropyrimidine-5-carboxylic acid ethyl ester, pubchem24010, ethyl2,4-dichloro-5-pyrimidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 221.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ethyl 2,4-dichloropyrimidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-531-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit