Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%

Click to view available options
Quantity:
250mg
1g
5g
51940-64-8
C7H6Cl2N2O2
221.037
MFCD09910281
SRJBDGLSCPDXBL-UHFFFAOYSA-N
ethyl 2,4-dichloro-5-pyrimidinecarboxylate,5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester,ethyl 2,4-dichloro-pyrimidine-5-carboxylate,2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester,2,4-dichloro-5-ethoxycarbonylpyrimidine,2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester,2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester,2,4-dichloropyrimidine-5-carboxylic acid ethyl ester,pubchem24010,ethyl2,4-dichloro-5-pyrimidinecarboxylate
104020
ethyl 2,4-dichloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)Cl
51940-64-8
C7H6Cl2N2O2
221.037
MFCD09910281
SRJBDGLSCPDXBL-UHFFFAOYSA-N
ethyl 2,4-dichloro-5-pyrimidinecarboxylate,5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester,ethyl 2,4-dichloro-pyrimidine-5-carboxylate,2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester,2,4-dichloro-5-ethoxycarbonylpyrimidine,2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester,2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester,2,4-dichloropyrimidine-5-carboxylic acid ethyl ester,pubchem24010,ethyl2,4-dichloro-5-pyrimidinecarboxylate
104020
ethyl 2,4-dichloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)Cl
Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color White
Melting Point 36°C to 37°C
Quantity 250mg
Beilstein 152173
Formula Weight 221.04
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Ethyl 2,4-dichloropyrimidine-5-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning