Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2,4-dioxo-5-methylhexanoate, 95%

Manufacturer:  Alfa Aesar™ B2573503

Quantity 1g

 View more versions of this product

Catalog No. AAB2573503


  Check Availability
Add to cart

Specifications

Description

Applications
Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

64195-85-3
186.21
2,4-Dioxo-5-methylhexanoic acid ethyl ester; Ethyl isobutyrylpyruvate
C9H14O4
MFCD07323638
Ethyl 2,4-dioxo-5-methylhexanoate

Specifications

64195-85-3
186.21
2,4-Dioxo-5-methylhexanoic acid ethyl ester; Ethyl isobutyrylpyruvate
Ethyl 2,4-dioxo-5-methylhexanoate
C9H14O4
MFCD07323638
1g
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit