Alfa Aesar™ Ethyl 2,4-dioxoheptanoate, 95%

Manufacturer: Alfa Aesar™ H3102706

Quantity 5g

 View more versions of this product

Catalog No. AAH3102706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36983-31-0
    Molecular Formula C9H14O4
    Formula Weight 186.21
    MDL Number MFCD08700411
    Synonym 2,4-Dioxoheptanoic acid ethyl ester
    Chemical Name or Material Ethyl 2,4-dioxoheptanoate

    Specifications

    Formula Weight 186.21
    Boiling Point 228° to 232°C
    Molecular Formula C9H14O4
    MDL Number MFCD08700411
    CAS 36983-31-0
    Synonym 2,4-Dioxoheptanoic acid ethyl ester
    Quantity 5g
    Chemical Name or Material Ethyl 2,4-dioxoheptanoate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit