Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
1g
10g
4815-24-1
C9H13NO2S
199.27
JYSDXODDWAQWJR-UHFFFAOYSA-N
ethyl 2-amino-4,5-dimethyl3-thenoate,ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate,2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester,2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester,3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester,2-amino-3-carbethoxy-4,5-dimethylthiophene,2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene,ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate,ethyl2-amino-4,5-dimethylthiophene-3-carboxylate,ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
78536
ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
CCOC(=O)C1=C(SC(=C1C)C)N
4815-24-1
C9H13NO2S
199.27
JYSDXODDWAQWJR-UHFFFAOYSA-N
ethyl 2-amino-4,5-dimethyl3-thenoate,ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate,2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester,2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester,3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester,2-amino-3-carbethoxy-4,5-dimethylthiophene,2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene,ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate,ethyl2-amino-4,5-dimethylthiophene-3-carboxylate,ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
78536
ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
CCOC(=O)C1=C(SC(=C1C)C)N
NMR Consistent with structure (1H−NMR)
Melting Point 90.0°C to 91.0°C
Infrared Spectrum Authentic
Quantity 10g
Assay Percent Range 96% min. (GC)
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 225-387-5