Learn More
Alfa Aesar™ Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%
Manufacturer:  Alfa Aesar™ A1079003
Quantity 1g
 View more versions of this product
Catalog No. AAA1079003
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
43088-42-2
185.25
1366073
Ethyl 2-amino-4-methylthiophene-3-carboxylate
C8H11NO2S
MFCD00051669
2-Amino-4-methylthiophene-3-carboxylic acid ethyl ester
Specifications
43088-42-2
185.25
1366073
79° to 83°C
Ethyl 2-amino-4-methylthiophene-3-carboxylate
C8H11NO2S
MFCD00051669
2-Amino-4-methylthiophene-3-carboxylic acid ethyl ester
1g
≥98%