Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-amino-5-iodobenzoate, 98+%

Click to view available options
Quantity:
25g
100g
268568-11-2
C9H10INO2
291.088
MFCD04038940
FPCLHSGOJPEKKE-UHFFFAOYSA-N
ethyl2-amino-5-iodobenzoate,2-amino-5-iodobenzoic acid ethyl ester,2-amino-5-iodo-benzoic acid ethyl ester,acmc-1cksm,ethyl 2-amino-5-iodo-benzoate,ethyl 5-iodoanthranilate
12012179
ethyl 2-amino-5-iodobenzoate
CCOC(=O)C1=C(C=CC(=C1)I)N
268568-11-2
C9H10INO2
291.088
MFCD04038940
FPCLHSGOJPEKKE-UHFFFAOYSA-N
ethyl2-amino-5-iodobenzoate,2-amino-5-iodobenzoic acid ethyl ester,2-amino-5-iodo-benzoic acid ethyl ester,acmc-1cksm,ethyl 2-amino-5-iodo-benzoate,ethyl 5-iodoanthranilate
12012179
ethyl 2-amino-5-iodobenzoate
CCOC(=O)C1=C(C=CC(=C1)I)N
Melting Point 69°C to 71°C
Quantity 100g
Formula Weight 291.09
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-amino-5-iodobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning