Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-aminobenzothiazole-6-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
25g
50850-93-6
C10H10N2O2S
222.262
MFCD00102724
VYJSGJXWKSDUSG-UHFFFAOYSA-N
ethyl 2-aminobenzo d thiazole-6-carboxylate,ethyl 2-amino-benzothiazole-6-carboxylate,ethyl 2-aminobenzothiazole-6-carboxylate,2-amino-benzothiazole-6-carboxylic acid ethyl ester,6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester,2-amino-6-ethoxycarbonylbenzothiazole,ethyl2-amino-benzothiazole-6-carboxylate,2-aminobenzothiazole-6-carboxylic acid ethyl ester,2-amino benzo thiazole-6-carboxylic acid ethyl ester,3kid
601008
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
50850-93-6
C10H10N2O2S
222.262
MFCD00102724
VYJSGJXWKSDUSG-UHFFFAOYSA-N
ethyl 2-aminobenzo d thiazole-6-carboxylate,ethyl 2-amino-benzothiazole-6-carboxylate,ethyl 2-aminobenzothiazole-6-carboxylate,2-amino-benzothiazole-6-carboxylic acid ethyl ester,6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester,2-amino-6-ethoxycarbonylbenzothiazole,ethyl2-amino-benzothiazole-6-carboxylate,2-aminobenzothiazole-6-carboxylic acid ethyl ester,2-amino benzo thiazole-6-carboxylic acid ethyl ester,3kid
601008
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
Quantity 5g
Melting Point 241°C to 245°C
Beilstein 185097
Formula Weight 222.26
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Ethyl 2-aminobenzothiazole-6-carboxylate