Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-aminonicotinate, 98%

Click to view available options
Quantity:
5g
25g
100g
13362-26-0
C8H10N2O2
166.18
MFCD01569452
KIAGEDYOPMHRRB-UHFFFAOYSA-N
ethyl 2-aminonicotinate,2-amino-nicotinic acid ethyl ester,ethyl2-aminonicotinate,2-aminonicotinic acid ethyl ester,3-pyridinecarboxylic acid, 2-amino-, ethyl ester,2-amino nicotinic acid ethyl ester,2-aminopyridine-3-carboxylic acid ethyl ester,pubchem12921,rarechem al bi 1234,buttpark 23\07-65
12433260
ethyl 2-aminopyridine-3-carboxylate
CCOC(=O)C1=C(N=CC=C1)N
13362-26-0
C8H10N2O2
166.18
MFCD01569452
KIAGEDYOPMHRRB-UHFFFAOYSA-N
ethyl 2-aminonicotinate,2-amino-nicotinic acid ethyl ester,ethyl2-aminonicotinate,2-aminonicotinic acid ethyl ester,3-pyridinecarboxylic acid, 2-amino-, ethyl ester,2-amino nicotinic acid ethyl ester,2-aminopyridine-3-carboxylic acid ethyl ester,pubchem12921,rarechem al bi 1234,buttpark 23\07-65
12433260
ethyl 2-aminopyridine-3-carboxylate
CCOC(=O)C1=C(N=CC=C1)N
Quantity 100g
Melting Point 90°C to 96°C (literature)
Beilstein 124680
Formula Weight 166.18
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Ethyl 2-aminonicotinate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning