Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-bromo-4-methylthiazole-5-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5183306

Quantity 5g

 View more versions of this product

Catalog No. AAH5183306


  Check Availability
Add to cart

Specifications

Description

Applications
It finds its application in the preparation of thiazole-4- and -5-carboxylates, and an infrared study of their rotational isomers and in the discovery of thiazolylpyridinone SCD1 inhibitors with preferential liver distribution and reduced mechanism-based adverse effects.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

22900-83-0
250.12
Ethyl 2-bromo-4-methylthiazole-5-carboxylate
C7H8BrNO2S
MFCD03791227

Specifications

22900-83-0
250.12
5g
97%
C7H8BrNO2S
MFCD03791227
Ethyl 2-bromo-4-methylthiazole-5-carboxylate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit