Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250mg
1g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
Solubility
Sparingly soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
Solubility
Sparingly soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Beilstein | 4426004 |
Quantity | 250mg |
Solubility Information | Sparingly soluble in water. |
Formula Weight | 274.13 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only