Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-bromohexanoate, 99+%

Manufacturer:  Alfa Aesar™ B2527314

Quantity 25g

 View more versions of this product

Catalog No. AAB2527314


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

615-96-3
223.12
UN3265
Ethyl 2-bromohexanoate
CH3(CH2)3CHBrCO2CH2CH3
MFCD00000162
2-Bromohexanoic acid ethyl ester

Specifications

615-96-3
CH3(CH2)3CHBrCO2CH2CH3
Pungent
MFCD00000162
2-Bromohexanoic acid ethyl ester
≥99%
213° to 215°C
25g
223.12
UN3265
Ethyl 2-bromohexanoate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit