Alfa Aesar™ Ethyl 2-bromohexanoate, 99+%

Manufacturer: Alfa Aesar™ B2527322

Quantity 100g

 View more versions of this product

Catalog No. AAB2527322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 615-96-3
    Molecular Formula CH3(CH2)3CHBrCO2CH2CH3
    Formula Weight 223.12
    MDL Number MFCD00000162
    UN Number UN3265
    Synonym 2-Bromohexanoic acid ethyl ester
    Chemical Name or Material Ethyl 2-bromohexanoate

    Specifications

    Formula Weight 223.12
    TSCA TSCA
    Boiling Point 213° to 215°C
    Molecular Formula CH3(CH2)3CHBrCO2CH2CH3
    Quantity 100g
    Odor Pungent
    MDL Number MFCD00000162
    CAS 615-96-3
    UN Number UN3265
    Synonym 2-Bromohexanoic acid ethyl ester
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Ethyl 2-bromohexanoate
    Assay ≥99%

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit