Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-bromoisobutyrate, 98+%

Click to view available options
Quantity:
50g
250g
600-00-0
C6H11BrO2
195.056
MFCD00000123
IOLQWGVDEFWYNP-UHFFFAOYSA-N
ethyl 2-bromoisobutyrate,ethyl 2-bromo-2-methylpropionate,ethyl alpha-bromoisobutyrate,ethyl .alpha.-bromoisobutyrate,propanoic acid, 2-bromo-2-methyl-, ethyl ester,2-bromo-2-methylpropanoic acid, ethyl ester,ethyl-2-bromoisobutyrate,propanoic acid, bromo-2-methyl-, ethyl ester,ethyl 2-bromo-2-methyl-propanoate,propionic acid, 2-bromo-2-methyl-, ethyl ester
11745
ethyl 2-bromo-2-methylpropanoate
CCOC(=O)C(C)(C)Br
600-00-0
C6H11BrO2
195.056
MFCD00000123
IOLQWGVDEFWYNP-UHFFFAOYSA-N
ethyl 2-bromoisobutyrate,ethyl 2-bromo-2-methylpropionate,ethyl alpha-bromoisobutyrate,ethyl .alpha.-bromoisobutyrate,propanoic acid, 2-bromo-2-methyl-, ethyl ester,2-bromo-2-methylpropanoic acid, ethyl ester,ethyl-2-bromoisobutyrate,propanoic acid, bromo-2-methyl-, ethyl ester,ethyl 2-bromo-2-methyl-propanoate,propionic acid, 2-bromo-2-methyl-, ethyl ester
11745
ethyl 2-bromo-2-methylpropanoate
CCOC(=O)C(C)(C)Br
Applications
Ethyl 2-bromoisobutyrate is used as an Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents, acids, bases, reducing agents.
Flash Point 57°C (135°F)
Boiling Point 162°C to 164°C
Quantity 250g
UN Number UN2924
Beilstein 1098947
Formula Weight 195.06
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromoisobutyrate

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA