Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-(bromomethyl)acrylate, 97%

Click to view available options
Quantity:
250mg
1g
5g
17435-72-2
C6H9BrO2
193.04
MFCD00031518
MTCMFVTVXAOHNQ-UHFFFAOYSA-N
ethyl 2-bromomethyl acrylate,ethyl 2-bromomethyl prop-2-enoate,2-bromomethyl acrylic acid ethyl ester,ethyl alpha-bromomethyl acrylate,2-propenoic acid, 2-bromomethyl-, ethyl ester,2-bromomethyl-acrylic acid ethyl ester,ethyl-2-bromomethylacrylate,ethyl 2-bromomethylacrylate,acmc-1bo78,ksc181c2b
310620
ethyl 2-(bromomethyl)prop-2-enoate
CCOC(=O)C(=C)CBr
17435-72-2
C6H9BrO2
193.04
MFCD00031518
MTCMFVTVXAOHNQ-UHFFFAOYSA-N
ethyl 2-bromomethyl acrylate,ethyl 2-bromomethyl prop-2-enoate,2-bromomethyl acrylic acid ethyl ester,ethyl alpha-bromomethyl acrylate,2-propenoic acid, 2-bromomethyl-, ethyl ester,2-bromomethyl-acrylic acid ethyl ester,ethyl-2-bromomethylacrylate,ethyl 2-bromomethylacrylate,acmc-1bo78,ksc181c2b
310620
ethyl 2-(bromomethyl)prop-2-enoate
CCOC(=O)C(=C)CBr
Quantity 250mg
Color Colorless to Yellow
Density 1.398g/mL
Boiling Point 38°C (0.8mmHg)
Refractive Index 1.4790
Beilstein 970108
Formula Weight 193.04
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 2-(bromomethyl)acrylate

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C